3-[2-(2-Bromo-4-fluorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050210-500MG 050210-2.5G
101890-740EA 364.62 USD
101890-740 101890-742
3-[2-(2-Bromo-4-fluorophenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Bromo-4-fluorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈BrClFNO
MDL Number: MFCD13560611
CAS Number: 1220017-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR