4-[2-(2-Bromo-4-fluorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050209-500MG 050209-2.5G
101890-736EA 364.62 USD
101890-736 101890-738
4-[2-(2-Bromo-4-fluorophenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Bromo-4-fluorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈BrClFNO
MDL Number: MFCD13560610
CAS Number: 1219982-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR