3-[2-(3,4-Difluorophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050206-500MG
050206-2.5G
101890-724EA
364.62
USD
101890-724
101890-726
3-[2-(3,4-Difluorophenoxy)ethyl]piperidine hydrochloride
3-[2-(3,4-Difluorophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈ClF₂NO |
MDL Number:
MFCD13560607 CAS Number: 1220016-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...