4-[2-(3,4-Difluorophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050205-500MG
050205-2.5G
101890-720EA
364.62
USD
101890-720
101890-722
4-[2-(3,4-Difluorophenoxy)ethyl]piperidine hydrochloride
4-[2-(3,4-Difluorophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈ClF₂NO |
MDL Number:
MFCD13560606 CAS Number: 1057343-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...