3-[(2,4-Difluorophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050204-500MG 050204-2.5G
101890-716EA 364.62 USD
101890-716 101890-718
3-[(2,4-Difluorophenoxy)methyl]pyrrolidine hydrochloride
3-[(2,4-Difluorophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClF₂NO
MDL Number: MFCD13560605
CAS Number: 1220032-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR