4-[2-(2,4-Difluorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050201-500MG 050201-2.5G
101890-704EA 364.62 USD
101890-704 101890-706
4-[2-(2,4-Difluorophenoxy)ethyl]piperidine hydrochloride
4-[2-(2,4-Difluorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClF₂NO
MDL Number: MFCD13560602
CAS Number: 1219964-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR