3-[(3-Methyl-4-nitrophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050200-500MG 050200-2.5G
101890-700EA 364.62 USD
101890-700 101890-702
3-[(3-Methyl-4-nitrophenoxy)methyl]pyrrolidine hydrochloride
3-[(3-Methyl-4-nitrophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₃
MDL Number: MFCD13560601
CAS Number: 1220029-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR