2-[2-(3-Methyl-4-nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050199-500MG 050199-2.5G
101890-696EA 364.62 USD
101890-696 101890-698
2-[2-(3-Methyl-4-nitrophenoxy)ethyl]piperidine hydrochloride
2-[2-(3-Methyl-4-nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O₃
MDL Number: MFCD13560600
CAS Number: 1219964-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR