3-[2-(3-Methyl-4-nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050198-500MG 050198-2.5G
101890-692EA 364.62 USD
101890-692 101890-694
3-[2-(3-Methyl-4-nitrophenoxy)ethyl]piperidine hydrochloride
3-[2-(3-Methyl-4-nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O₃
MDL Number: MFCD13560599
CAS Number: 1220038-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR