3-[(4-Nitrophenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
050196-500MG
050196-2.5G
101890-684EA
364.62
USD
101890-684
101890-686
3-[(4-Nitrophenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Nitrophenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₂O₃ |
MDL Number:
MFCD13560597 CAS Number: 1219982-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...