2-[2-(4-Nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050195-500MG 050195-2.5G
101890-680EA 364.62 USD
101890-680 101890-682
2-[2-(4-Nitrophenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₃
MDL Number: MFCD13560596
CAS Number: 1220032-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR