3-[2-(4-Nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050194-500MG 050194-2.5G
101890-676EA 364.62 USD
101890-676 101890-678
3-[2-(4-Nitrophenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₃
MDL Number: MFCD13560595
CAS Number: 1220032-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR