4-[2-(4-Nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050193-500MG 050193-2.5G
101890-672EA 364.62 USD
101890-672 101890-674
4-[2-(4-Nitrophenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₃
MDL Number: MFCD13560594
CAS Number: 1220037-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR