3-[(3-Nitrophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050192-500MG 050192-2.5G
101890-668EA 364.62 USD
101890-668 101890-670
3-[(3-Nitrophenoxy)methyl]pyrrolidine hydrochloride
3-[(3-Nitrophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₃
MDL Number: MFCD13560593
CAS Number: 1219982-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR