3-[2-(3-Nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050190-500MG 050190-2.5G
101890-660EA 364.62 USD
101890-660 101890-662
3-[2-(3-Nitrophenoxy)ethyl]piperidine hydrochloride
3-[2-(3-Nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₃
MDL Number: MFCD13560591
CAS Number: 1220032-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR