3-[(4-Methyl-2-nitrophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050188-500MG 050188-2.5G
101890-652EA 364.62 USD
101890-652 101890-654
3-[(4-Methyl-2-nitrophenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Methyl-2-nitrophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₃
MDL Number: MFCD13560589
CAS Number: 1220032-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR