3-[2-(4-Methyl-2-nitrophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050186-500MG
050186-2.5G
101890-644EA
364.62
USD
101890-644
101890-646
3-[2-(4-Methyl-2-nitrophenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Methyl-2-nitrophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁ClN₂O₃ |
MDL Number:
MFCD13560587 CAS Number: 1220032-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...