8-(3-Pyrrolidinylmethoxy)quinoline hydrochloride

Supplier: Matrix Scientific

050184-500MG 050184-2.5G
101890-636EA 166.15 USD
101890-636 101890-638
8-(3-Pyrrolidinylmethoxy)quinoline hydrochloride
8-(3-Pyrrolidinylmethoxy)quinoline hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₇ClN₂O
MDL Number: MFCD13560585
CAS Number: 1219982-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR