8-[2-(3-Piperidinyl)ethoxy]quinoline hydrochloride

Supplier: Matrix Scientific

050182-500MG 050182-2.5G
101890-628EA 364.62 USD
101890-628 101890-630
8-[2-(3-Piperidinyl)ethoxy]quinoline hydrochloride
8-[2-(3-Piperidinyl)ethoxy]quinoline hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₁ClN₂O
MDL Number: MFCD13560583
CAS Number: 1220029-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR