8-[2-(4-Piperidinyl)ethoxy]quinoline hydrochloride

Supplier: Matrix Scientific

050181-500MG 050181-2.5G
101890-624EA 364.62 USD
101890-624 101890-626
8-[2-(4-Piperidinyl)ethoxy]quinoline hydrochloride
8-[2-(4-Piperidinyl)ethoxy]quinoline hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₁ClN₂O
MDL Number: MFCD13560582
CAS Number: 1219982-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR