3-(3-Pyrrolidinylmethoxy)pyridine hydrochloride

Supplier: Matrix Scientific

050180-500MG 050180-2.5G
101890-620EA 364.62 USD
101890-620 101890-622
3-(3-Pyrrolidinylmethoxy)pyridine hydrochloride
3-(3-Pyrrolidinylmethoxy)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD13560581
CAS Number: 1220016-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR