4-Chloro-1-naphthyl-3-pyrrolidinylmethyl ether hydrochloride
Supplier: Matrix Scientific
050176-500MG
050176-2.5G
101890-604EA
364.62
USD
101890-604
101890-606
4-Chloro-1-naphthyl-3-pyrrolidinylmethyl ether hydrochloride
4-Chloro-1-naphthyl-3-pyrrolidinylmethyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₅H₁₇Cl₂NO |
MDL Number:
MFCD13560577 CAS Number: 1220037-78-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...