2-{2-[(4-Chloro-1-naphthyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050175-500MG 050175-2.5G
101890-600EA 364.62 USD
101890-600 101890-602
2-{2-[(4-Chloro-1-naphthyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(4-Chloro-1-naphthyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₁Cl₂NO
MDL Number: MFCD13560576
CAS Number: 1220032-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR