3-{2-[(4-Chloro-1-naphthyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050174-500MG 050174-2.5G
101890-596EA 364.62 USD
101890-596 101890-598
3-{2-[(4-Chloro-1-naphthyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(4-Chloro-1-naphthyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₁Cl₂NO
MDL Number: MFCD13560575
CAS Number: 1220029-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR