2-[2-(4-Propylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050171-500MG
050171-2.5G
101890-584EA
364.62
USD
101890-584
101890-586
2-[2-(4-Propylphenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Propylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₆ClNO |
MDL Number:
MFCD13560572 CAS Number: 1220037-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...