3-[2-(4-Propylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050170-500MG 050170-2.5G
101890-580EA 364.62 USD
101890-580 101890-582
3-[2-(4-Propylphenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Propylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560571
CAS Number: 1219972-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR