4-[2-(4-Propylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050169-500MG 050169-2.5G
101890-576EA 364.62 USD
101890-576 101890-578
4-[2-(4-Propylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Propylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560570
CAS Number: 1219982-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR