3-[(4-Ethylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050168-500MG 050168-2.5G
101890-572EA 364.62 USD
101890-572 101890-574
3-[(4-Ethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Ethylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD13560569
CAS Number: 1219982-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR