2-[2-(4-Ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050167-500MG 050167-2.5G
101890-568EA 364.62 USD
101890-568 101890-570
2-[2-(4-Ethylphenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560568
CAS Number: 1220016-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR