3-[2-(4-Ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050166-500MG 050166-2.5G
101890-564EA 364.62 USD
101890-564 101890-566
3-[2-(4-Ethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560567
CAS Number: 1220032-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR