4-[2-(4-Ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050165-500MG 050165-2.5G
101890-560EA 364.62 USD
101890-560 101890-562
4-[2-(4-Ethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560566
CAS Number: 1219964-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR