3-[(4-Benzylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050164-500MG 050164-2.5G
101890-556EA 364.62 USD
101890-556 101890-558
3-[(4-Benzylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Benzylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MDL Number: MFCD13560565
CAS Number: 1220016-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR