2-[2-(4-Benzylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050163-500MG
050163-2.5G
101890-552EA
364.62
USD
101890-552
101890-554
2-[2-(4-Benzylphenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Benzylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₂₀H₂₆ClNO |
MDL Number:
MFCD13560564 CAS Number: 152009-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...