3-[(2-Methoxy-4-methylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050160-500MG 050160-2.5G
101890-540EA 364.62 USD
101890-540 101890-542
3-[(2-Methoxy-4-methylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Methoxy-4-methylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD13560561
CAS Number: 1219964-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR