4-[2-(2-Methoxy-4-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050157-500MG 050157-2.5G
101890-528EA 364.62 USD
101890-528 101890-530
4-[2-(2-Methoxy-4-methylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Methoxy-4-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560558
CAS Number: 1220029-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR