2-{2-[4-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050155-500MG 050155-2.5G
101890-520EA 364.62 USD
101890-520 101890-522
2-{2-[4-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride
2-{2-[4-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560556
CAS Number: 1220016-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR