3-{2-[4-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050154-500MG 050154-2.5G
101890-516EA 364.62 USD
101890-516 101890-518
3-{2-[4-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[4-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560555
CAS Number: 1219982-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR