3-[(4-Isopropylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050152-500MG 050152-2.5G
101890-508EA 364.62 USD
101890-508 101890-510
3-[(4-Isopropylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Isopropylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD13560553
CAS Number: 1220029-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR