2-[2-(4-Isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050151-500MG 050151-2.5G
101890-504EA 364.62 USD
101890-504 101890-506
2-[2-(4-Isopropylphenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560552
CAS Number: 1220032-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR