4-[2-(4-Isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050149-500MG 050149-2.5G
101890-496EA 364.62 USD
101890-496 101890-498
4-[2-(4-Isopropylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560550
CAS Number: 1219982-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR