3-{[4-(tert-Pentyl)phenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

050148-500MG 050148-2.5G
101890-492EA 364.62 USD
101890-492 101890-494
3-{[4-(tert-Pentyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[4-(tert-Pentyl)phenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560549
CAS Number: 1220032-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR