2-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050143-500MG 050143-2.5G
101890-472EA 364.62 USD
101890-472 101890-474
2-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethyl}piperidine hydrochloride
2-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₂₁H₃₆ClNO
MDL Number: MFCD13560544
CAS Number: 1219982-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR