4-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050141-500MG 050141-2.5G
101890-464EA 364.62 USD
101890-464 101890-466
4-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₂₁H₃₆ClNO
MDL Number: MFCD13560542
CAS Number: 1219972-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR