3-{[4-(1-Methyl-1-phenylethyl)phenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

050139-500MG 050139-2.5G
101890-456EA 364.62 USD
101890-456 101890-458
3-{[4-(1-Methyl-1-phenylethyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[4-(1-Methyl-1-phenylethyl)phenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₆ClNO
MDL Number: MFCD13560540
CAS Number: 1220032-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR