2-{2-[4-(1-Methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride
Supplier: Matrix Scientific
050138-500MG
050138-2.5G
101890-452EA
364.62
USD
101890-452
101890-454
2-{2-[4-(1-Methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride
Hazard: Irritant
Formula:
C₂₂H₃₀ClNO |
MDL Number:
MFCD00944210 CAS Number: 174775-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...