3-{2-[4-(1-Methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050137-500MG 050137-2.5G
101890-448EA 186.15 USD
101890-448 101890-450
3-{2-[4-(1-Methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[4-(1-Methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₂₂H₃₀ClNO
MDL Number: MFCD13560539
CAS Number: 1219963-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR