4-{2-[4-(1-Methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050136-500MG 050136-2.5G
101890-444EA 364.62 USD
101890-444 101890-446
4-{2-[4-(1-Methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[4-(1-Methyl-1-phenylethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₂₂H₃₀ClNO
MDL Number: MFCD13560538
CAS Number: 1220020-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR