4-{2-[4-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050132-500MG 050132-2.5G
101890-428EA 364.62 USD
101890-428 101890-430
4-{2-[4-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[4-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClF₃NO
MDL Number: MFCD13560534
CAS Number: 1219972-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR