Methyl-4-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride

Supplier: Matrix Scientific

050129-500MG 050129-2.5G
101890-416EA 364.62 USD
101890-416 101890-418
Methyl-4-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride
Methyl-4-[2-(3-piperidinyl)ethoxy]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13560531
CAS Number: 1220032-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR