Methyl-4-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride
Supplier: Matrix Scientific
050128-500MG
050128-2.5G
101890-412EA
364.62
USD
101890-412
101890-414
Methyl-4-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride
Methyl-4-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₂ClNO₃ |
MDL Number:
MFCD13560530 CAS Number: 1112052-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...